Learn More
Simport™ Scientific Histosette™ I Tissue Processing/Embedding Cassette
Catalog No. 22045054 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045-054 Aqua
22-045-044 White
22-045-045 Pink
22-045-046 Green
22-045-047 Yellow
22-045-048 Blue
22-045-049 Peach
22-045-050 Tan
22-045-051 Gray
22-045-052 Lilac
22-045-053 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-045-054 Supplier Simport™ Scientific Supplier No. M49012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ I Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
  • Made of acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to the chemical action of most histological solvents
  • 30° angle anterior writing area
  • One-piece construction with frangible hinges for easy lid removal
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary - cassettes will always relock securely
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Closure Included Yes
Color Aqua
Material Acetal
Product Type Tissue Cassette
For in vitro diagnostic (IVD) use only.