Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Catalog No. 22045065 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045-065 Aqua
22-045-055 White
22-045-056 Pink
22-045-057 Green
22-045-058 Yellow
22-045-059 Blue
22-045-060 Peach
22-045-061 Tan
22-045-062 Gray
22-045-063 Lilac
22-045-064 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-045-065 Supplier Simport™ Scientific Supplier No. M49212
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
  • Made of acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to the chemical action of most histological solvents
  • 45° angle anterior writing area
  • One-piece integral lid eliminates need for separate steel lids
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary - cassettes will always relock securely
  • Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Closure Included Yes
Color Aqua
Material Acetal
Product Type Tissue Cassette
For in vitro diagnostic (IVD) use only.