Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped)
Catalog No. 22038508 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Green
Orange
Peach
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-508 Yellow
22-038-045 White
22-038-047 Green
22-038-049 Blue
22-038-050 Peach
22-038-054 Orange
22-038-055 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-038-508 Supplier Simport™ Scientific Supplier No. M4925T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped). Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Open and close as often as necessary - cassettes will always relock securely
  • Back mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Yellow
Material Acetal