Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HisTrap™ FF Columns: 5 x 5mL
GSA_VA

Catalog No. 45000326 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-326 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-326 Supplier Cytiva Supplier No. 17525501
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Optimize purifications of histidine-tagged proteins directly from homogenized, unclarified cells HisTrap™ FF Columns: 5 x 5mL are prepacked with precharged Ni Sepharose™ 6 Fast Flow media.

  • Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
  • Column material: polypropylene
  • Bed dimensions: 7 × 25mm
  • Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
  • Compatible with wide range of reducing agents, detergents, denaturants, and other additives
  • Simple operation with syringe, perstaltic pump, or chromatographic system
  • Columns may be connected in series to increase capacity
Medium
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.2 in.
Diameter (Metric) Inner 7 mm
Description HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).
Flow Rate <4 mL/min.
Material Polypropylene
pH Range 3 to 12
Support Type 6% Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Quantity 5 x 5 mL
Type Pre-packed Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.