Learn More
Thermco Products HiTemp Data Logger
Catalog No. 04500355
Change view
Click to view available options
Display Type:
No Display
Catalog No. Display Type
04-500-355 No Display
1 options
Catalog No. 04-500-355 Supplier Thermco Products Supplier No. ACCDHL
Add to Cart
Add to Cart
HotLog Digital Datalogger for autoclave upto 140°C, internal sensor, waterproof and pressure-tight up to 10 bar.
  • This high-temperature data logger is designed for precise monitoring in sterilization, pasteurization, and applications up to 140°C, making it ideal for food, pharmaceutical, and laboratory sectors.
  • Features a robust, food-safe AISI 316L stainless steel housing that is waterproof and pressure-tight up to 10 bar (IP68).
  • Offers a wide measuring range from -40°C to 140°C, high accuracy of ±0.25°C in the calibrated range, and 0.01°C resolution.
  • Provides a large 60,000-record memory, adjustable recording intervals from 1 sec. to 24 hr., and MultiLog capability for up to 500 recordings.
  • Automatic F0 and L value calculations ensure compliance with food safety standards.
  • User-replaceable 3.6 V LiSoCl2 battery supports up to 5,000,000 measurements or 3 Years of standby operation.
  • Data is managed using LOG-Connect software and a USB readout unit, making it suitable for quality monitoring in production and sterilization processes.
  • Compact dimensions of 23 x 40 mm and a weight of 75 g support easy integration into various applications.
Specifications
Type Datalogger
Display Type No Display
Alarm Types None