Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hitfinder plate no. 072 for HTS, 80 compounds, 1 micromol each, Thermo Scientific™

Catalog No. HF01072
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
HF01072 1 Ea. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HF01072 Supplier Thermo Scientific Chemicals Supplier No. HF01072

Specifications

Chemical Name or Material Hitfinder plate no 072 for HTS, 80 compounds 1micromol each
Packaging Amber Glass Bottle
Quantity 1 Ea.
Formula Weight 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.