Learn More
Hitfinder plate no. 097 for HTS, 80 compounds, 1 micromol each, Thermo Scientific™
Specifications
Specifications
| Chemical Name or Material | Hitfinder plate no 097 for HTS, 80 compounds 1micromol each |
| Packaging | Amber Glass Bottle |
| Quantity | 1 Ea. |
| Formula Weight | 0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.