Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ HiTrap™ Antibody Protein G Package

Catalog No. 01257023 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-257-023 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-023 Supplier Cytiva™ Supplier No. 29058806
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein G package enables convenient purification of most species of antibodies, including rat.

The HiTrap™ Protein G HP column enclosed is recommended for most species of antibodies. A HiTrap™ Desalting column is also provided for quick pH neutralization of the eluted antibodies.

  • Contains HiTrap™ Protein G HP (1 x 1 mL) HiTrap™ Desalting (1 x 5 mL)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antibody Purification Kit
Product Line HiTrap
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.