Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Blue HP Columns

Catalog No. 45000063 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Bed Volume:
1mL
5mL
Diameter (Metric) Inner:
0.7cm
1.6cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate Bed Volume Diameter (Metric) Inner
45-000-063 5 x 1mL <4 mL/min. 1mL 0.7cm
45-000-064 1 x 5mL <20 mL/min. 5mL 1.6cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-063 Supplier Cytiva Supplier No. 17041201
Add to Cart
Add to Cart

For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins

  • Prepacked with Blue Sepharose High Performance media
  • Cibacron Blue F3G-A, ligand, is covalently attached to the 6% agarose medium
  • Stable over pH range 4 to 12 for use and 3 to 13 for cleaning

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 1mL
Flow Rate <4 mL/min.
Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 1mL
Product Type Blue HP Columns
Diameter (Metric) Inner 0.7cm
For Use With (Application) Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type 4mg Cibacron blue F3G-A/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 4 to 12
Support Type 6% Highly Cross-linked Agarose
Sample Type Albumin, Interferon, Nucleotide-dependent Enzymes, a2-macroglobulin, Coagulation Factors and Related Proteins
Content And Storage 4° to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.