Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Butyl-S FF Columns

Catalog No. 45000174 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Diameter (English) Inner:
0.275 in.
Flow Rate:
< 1 mL/min
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Flow Rate
45-000-174 5 x 1 mL 0.275 in. < 1 mL/min
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-174 Supplier Cytiva Supplier No. 17097813
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for purification of relatively strong hydrophilic molecules at low salt concentrations

  • Useful for removal of strong hydrophobic contaminants, such as lipoproteins, lipids, pigments and more from biological samples
  • Used successfully for large-scale purification of recombinant Hepatitis B virus surface antigen (rec-HBsAg) from CHO cells
  • Robust production with high reproducibility

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.275 in.
Diameter (Metric) Inner 7 mm
Product Type HiTrap Butyl-S FF column
Flow Rate < 1 mL/min
For Use With (Application) For Screening of Selectivity, Binding and Elution Conditions, as Well as Small Scale Purifications
Quantity 5 x 1 mL
Ligand Type Butyl-S
Material Polypropylene
Max. Pressure 5 bar [0.5 MPa] (70 psi)
Particle Size 45 to 165 μm
pH Range 3 to 13
Support Type 6% Cross-linked Agarose
Storage Requirements 4°C to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.