Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ Adhere Columns

Catalog No. 45001439 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.30 in.
0.62 in.
Flow Rate:
<20 mL/min.
<4 mL/min.
Max. Pressure:
1.5 bar
44 bar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Flow Rate Max. Pressure
45-001-439 5 x 5mL 0.62 in. <20 mL/min. 44 bar
45-001-438 5 x 1mL 0.30 in. <4 mL/min. 1.5 bar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-439 Supplier Cytiva Supplier No. 28405846
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to remove contaminants in post-protein A purification

  • Prepacked with Capto adhere media, a strong anion exchanger with multimodal functionality
  • High capacity and productivity in flow-through mode: 0.09 to 0.12mmol Cl-/mL medium ionic capacity
  • Wide operational window of pH and conductivity: working pH range 3 to 12
  • Two-step process reduces time spent and operating costs

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto adhere column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 5 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type N-benzyl-n-methyl Ethanolamine
Max. Pressure 44 bar
pH Range 3 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Easy screening and convenient process development
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.