Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ Core 400

Catalog No. 09928115 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-115 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-928-115 Supplier Cytiva Supplier No. 17372411
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for intermediate purification and polishing of viruses and other large biomolecules

  • An excellent alternative to size‐exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacturing
  • Novel core bead technology and multimodal ligands provide dual functionality, size exclusion, and binding chromatography
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
  • High capacity and productivity in flowthrough mode
  • Wide operational window of pH and conductivity
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5/Pk.
Bed Volume 1 mL
For Use With (Application) Intermediate Purification, and Polishing of Viruses and Large Biomolecules
Max. Pressure 0.5 MPa (5 bar)
Stationary Phase Capto™ Core 400
Product Line HiTrap
Column Type Prepacked HiTrap™ Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.