Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ Core 700 Columns

Catalog No. 45002598 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-598 5 x 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-598 Supplier Cytiva Supplier No. 17548151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiTrap Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess medium for intermediate purification and polishing of viruses and other large biomolecules.

Capto Core 700 is a novel, layered bead chromatography medium consisting of a porous outer layer and ligand-activated core.

  • High capacity and productivity in flowthrough mode.
  • Wide operational window of pH and conductivity.
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification.
  • The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged.
  • These internalized ligands bind impurities strongly over a wide range of pH and salt concentration.
  • The pores of the bead outer layer prevent large targets (Mr > 700 000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type HiTrap Capto Core 700 multimodal column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Flow Rate <500cm/hr.
Material Polypropylene
Ligand Type Octylamine
Max. Pressure 5 bar
pH Range 2 to 14
Particle Size 90μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Intermediate purification and polishing of viruses and other large biomolecules
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.