Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap Capto MMC Columns

Catalog No. 45000004 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Flow Rate:
<4 mL/min.
300kPa at 600cm/hr.
Max. Pressure:
3 bar
44 bar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Flow Rate Max. Pressure
45-000-004 5 x 1mL 0.27 in. <4 mL/min. 44 bar
45-000-005 5 x 5mL 0.62 in. 300kPa at 600cm/hr. 3 bar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-004 Supplier Cytiva Supplier No. 11003273
Add to Cart
Add to Cart

HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.

  • High dynamic binding capacity at high conductivity.
  • Selectivity allows binding in the presence of salt.
  • High volume throughput.
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto MMC column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Max. Pressure 44 bar
pH Range 2 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Bind Proteins at the conductivity of the feed material and to solve specific purification challenges
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.