Learn More
Cytiva HiTrap™ Capto™ MMC Impres Multimodal Chromatography Column
GREENER_CHOICE
Catalog No. 45002502 Shop All Cytiva Products
Change view
Click to view available options
Particle Size:
40 μm
Catalog No. Particle Size
45-002-502 40 μm
1 options
Catalog No. 45-002-502 Supplier Cytiva Supplier No. 17371610
Add to Cart
Add to Cart
Ready-to-use column for screening and small-scale protein purifications using ion exchange chromatography (IEX)
  • High-resolution beads and ligand selectivity ensures high yields
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window
  • For screening of selectivity, binding, and elution conditions
  • Removes contaminants such as DNA, host cell proteins (HCP), leached protein A, and aggregates
  • Can be used in initial polishing step in MAb purification processes, as well as for polishing of antibody fragments such as domain antibodies
Avoid using oxidizing agents, cationic detergents.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC, MAbs Purification
Max. Pressure 300,000 psi
Particle Size 40 μm