Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap Capto S ImpAct Prepacked Columns

Catalog No. 45003203 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Flow Rate:
20 mL/min.
4 mL/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate
45-003-203 5 x 5mL 20 mL/min.
45-003-202 5 x 1mL 4 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-003-203 Supplier Cytiva Supplier No. 17371755
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).

  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution intermediate purification and polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
TRUSTED_SUSTAINABILITY

Specifications

Product Type HiTrap Capto S ImpAct Prepacked Column
Quantity 5 x 5mL
Flow Rate 20 mL/min.
Material High-flow agarose
pH Range 4 to 12
Particle Size 50μm
Column Type Ion Exchange Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.