Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Chelating HP Columns

Catalog No. 45000061 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
5 x 5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-061 1 x 5mL
45-000-060 5 x 1mL
45-000-062 5 x 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-000-061 Supplier Cytiva Supplier No. 17040901
Add to Cart
Add to Cart

Optimize histidine-tagged protein purifications when nickel ions are not the best choice. Cytiva Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.

  • Uses different kinds of metal ions
  • Binding capacity ∽12mg histidine-tagged protein/mL medium
  • Metal ion capacity ∽23μmol Cu2+/mL medium
  • Prepacked with Chelating Sepharose High Performance media
  • Iminoacetic acid coupled to a 6% agarose matrix by ether linkage

TRUSTED_SUSTAINABILITY

Specifications

Buffer 6M Guanidine Hydrochloride, 8M Urea
Product Type HiTrap chelating HP column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type Iminodiacetic Acid
Max. Pressure 3 bar
pH Range 3 to 13
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) IMAC and subsequent purification of Polyhistidine tagged Proteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.