Learn More
Cytiva HiTrap™/HiPrep™ Column Accessories
Catalog No. 45001426 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 Pack
8 Pack
Product Type:
Connector
Stop Plug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
45-001-426 8 Pack Connector
45-000-001 5 Pack Stop Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-426 Supplier Cytiva Supplier No. 28401081
Specifications
Product Type Connector
Quantity 8 Pack
Material Polypropylene
Column Type Column Accesory
For Use With (Application) HiTrap and HiPrep column (connecting column to AKTA™ design systems)