Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ IgM Purification HP Column
GSA_VA

Catalog No. 45000276 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-276 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-276 Supplier Cytiva Supplier No. 17511001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Complete instructions for use and method optimization, recommendations for sample preparation, and a variety of connectors for syringe, peristaltic pump, and chromatography system

Optimized for fast, efficient purification of monoclonal IgM from hybridoma cell culture

  • Prepacked with a thiophilic adsorption medium, 2-mercaptopyridine coupled to Sepharose™ High Performance medium
  • Recovery of IgM is high, with excellent activity retention; typical purity, >80%
  • Purification of human IgM possible; binding capacity: 5mg human IgM/mL medium
  • Designed for use with a syringe, peristaltic pump, or chromatography system

Order Info

Supplied in packs of five 1mL columns

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Binding Capacity 5 mg Human IgM/mL Medium
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 1.6 cm
Description HiTrap IgM Purification HP is a 1 mL column prepacked with Sepharose High Performance for fast and efficient purification of monoclonal IgM.
Flow Rate <4 mL/min.
Content And Storage 4 to 8°C, 20% Ethanol
Quantity 5 x 1 mL
For Use With (Application) Chromatography System, Manual
Type Pre-packed Column
Sample Volume 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.