Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ MabSelect™ Prepacked Columns

Catalog No. 45001463 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
5 x 5 mL
Product Line:
MabSelect
MabSelect™ Xtra
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Line
45-001-463 1 x 5 mL MabSelect
45-001-465 5 x 1 mL MabSelect™ Xtra
45-001-467 5 x 5 mL MabSelect™ Xtra
45-001-464 5 x 5 mL MabSelect
45-001-462 5 x 1 mL MabSelect
45-500-123 1 x 5 mL MabSelect™ Xtra
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-001-463 Supplier Cytiva Supplier No. 28408255
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use these columns in small-scale MAb purification in a continuous bioprocess. Cytiva Life Sciences™ HiTrap™ MabSelect™ Prepacked Columns for fast purification of monoclonal antibodies (Mabs) from large sample volumes

Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.

  • Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
  • High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system

MabSelect

  • Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
  • Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
MabSelect Xtra™
  • Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
  • Allows binding from samples with high expression levels of the monoclonal antibody
  • Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
MabSelect SuRe™ (Superior Resistance)
  • Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
  • Permits significant cost savings and extension of cycles in regular production

TRUSTED_SUSTAINABILITY

Specifications

Product Line MabSelect
Binding Capacity ∼30 mg/mL Human lgG at 2.4 min. residence time
Diameter (Metric) Inner Column 1.6 cm
Diameter (English) Inner Column 0.62 in.
Flow Rate <20 mL/min.
Volume (Metric) Bed 5 mL
For Use With (Application) For fast purification of monoclonal antibodies from large sample volumes, Protein Purification
Sample Volume 5 mL
Storage Buffer 20% Ethanol
Content And Storage 4°C to 30°C
Quantity 1 x 5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.