Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap MabSelect PrismA Column

Catalog No. 09928070 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-070 1 x 5 mL
09-928-068 1 x 1 mL
09-928-069 5 x 1 mL
09-928-071 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-928-070 Supplier Cytiva Supplier No. 17549853
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Protein A Column

These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Flow Rate <20 mL/min.
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Quantity 1 x 5 mL
For Use With (Application) Antibody affinity chromatography
Type Pre-packed Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.