Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ MabSelect™ SuRe™ LX Columns

Catalog No. p-7087581 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
5/Pk.
Volume (Metric) Bed:
1 mL
5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Volume (Metric) Bed
45-200-506 5/Pk. 1 mL
45-200-507 10/Pk. 1 mL
45-200-508 5/Pk. 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-200-506 Supplier Cytiva Supplier No. 29268402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polyclonal antibodies, monoclonal antibodies (mAb), and antibody fragments are usually purified by affinity chromatography. Resins containing an immobilized ligand (e.g., protein A, protein G, or protein L) are used to capture antibodies and antibody fragments.

  • Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes
  • Reduces the risk of product contamination and carryover
  • Improves performance and reduces overall costs
TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Column
Stationary Phase MabSelect SuRe LX
Flow Rate 4 mL/min. (Max)
Ligand Protein A
Particle Size 85 μm
Purification Target Human IgG
Volume (Metric) Bed 1 mL
For Use With (Application) Purification of Human IgG
Sample Volume 1 to 30 mg/Column
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.