Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ MabSelect SuRe™ PCC Columns

Catalog No. 09928120 Shop All Cytiva Products
Click to view available options
Bed Volume:
1 mL
5 mL

Prepacked with MabSelect SuRe™ PCC Protein A Resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC)

  • Suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions
  • Alkali-stabilized, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols
  • These protein A columns provide fast, reproducible, and easy separations in a convenient format
  • Highly concentrated elution pools for operating flexibility and small unit operations
  • Allows cost-effective cleaning with 0.1 to 0.5 M NaOH over hundreds of purification cycles
  • Efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result
  • Columns are optimally utilized when they are connected to liquid chromatography systems such as the ÄKTA systems
  • High binding capacity: ∼ 60 mg Human IgG/mL Resin at 2.4 min residence time
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chromatography Column
Bed Volume 1 mL
Diameter (Metric) Inner 7 mm
Quantity 5/Pk.
Height (Metric) 25 mm
Flow Rate <4 mL/min
Max. Pressure 5 bar (0.5 MPa)
Content And Storage Store at 2°C to 8°C, 20% Ethanol
Capacity ∼60 mg Human IgG/mL Resin
Column Type Affinity Column
For Use With (Application) Antibody Affinity Chromatography
Matrix MabSelect SuRe PCC Protein A Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.