Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ MabSelect SuRe™ PCC Columns

Catalog No. 09928121 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
09-928-121 5 mL
09-928-120 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-121 Supplier Cytiva Supplier No. 17549112

missing translation for 'validMainframeMsg'

Prepacked with MabSelect SuRe™ PCC Protein A Resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC)

  • Suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions
  • Alkali-stabilized, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols
  • These protein A columns provide fast, reproducible, and easy separations in a convenient format
  • Highly concentrated elution pools for operating flexibility and small unit operations
  • Allows cost-effective cleaning with 0.1 to 0.5 M NaOH over hundreds of purification cycles
  • Efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result
  • Columns are optimally utilized when they are connected to liquid chromatography systems such as the ÄKTA systems
  • High binding capacity: ∼ 60 mg Human IgG/mL Resin at 2.4 min residence time
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chromatography Column
Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Quantity 1/Pk.
Height (Metric) 25 mm
Flow Rate <20 mL/min
Max. Pressure 5 bar (0.5 MPa)
Content And Storage Store at 2°C to 8°C, 20% Ethanol
Capacity ∼60 mg Human IgG/mL Resin
Column Type Affinity Column
For Use With (Application) Antibody Affinity Chromatography
Matrix MabSelect SuRe PCC Protein A Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.