Learn More
Description
- Suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions
- Alkali-stabilized, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols
- These protein A columns provide fast, reproducible, and easy separations in a convenient format
- Highly concentrated elution pools for operating flexibility and small unit operations
- Allows cost-effective cleaning with 0.1 to 0.5 M NaOH over hundreds of purification cycles
- Efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result
- Columns are optimally utilized when they are connected to liquid chromatography systems such as the ÄKTA systems
- High binding capacity: ∼ 60 mg Human IgG/mL Resin at 2.4 min residence time
Specifications
Specifications
| Product Type | Chromatography Column |
| Bed Volume | 1 mL |
| Diameter (Metric) Inner | 7 mm |
| Quantity | 5/Pk. |
| Height (Metric) | 25 mm |
| Flow Rate | <4 mL/min |
| Max. Pressure | 5 bar (0.5 MPa) |
| Content And Storage | Store at 2°C to 8°C, 20% Ethanol |
| Capacity | ∼60 mg Human IgG/mL Resin |
| Column Type | Affinity Column |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.