Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Protein A HP Prepacked Columns
GSA_VA

Catalog No. 45000049 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 1 mL
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate
45-000-049 5 x 1 mL <4 mL/min.
45-000-052 5 x 5 mL <20 mL/min.
45-000-050 2 x 1 mL <4 mL/min.
45-000-051 1 x 5 mL <20 mL/min.
45-000-503 1 x 1 mL <4 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 45-000-049 Supplier Cytiva Supplier No. 17040201
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. Cytiva Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.

  • Prepacked with Protein A Sepharose High Performance media for purification and fractionation of IgG subclasses
  • For use with syringe, pump or chromatography system such as KTA design or FPLC System
  • Binds 20mg human IgG/mL medium
  • Stable over pH 3 to 9 for use and 2 to 10 for cleaning
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • 1 and 5mL columns

Media
  • Ligand: Native protein A
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 20 mg/mL IgG Medium
Flow Rate <4 mL/min.
For Use With (Application) Chromatography System, Manual
For Use With (Equipment) For syringe, peristaltic pump or a chromatography system
Content And Storage 4°C to 8°C, 20% Ethanol
Quantity 5 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.