Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap Protein L, 5 x 5ml
GSA_VA

Catalog No. 45002379 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 × 5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-379 5 × 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-379 Supplier Cytiva Supplier No. 17547855
Add to Cart
Add to Cart

Prepacked with Capto L. Cytiva HiTrap Protein L, 5 x 5ml facilitate easy protein purification. They are prefilled with Capto L, a BioProcess chromatography medium that combines a rigid, high-flow agarose matrix with the immunoglobulin-binding recombinant protein, L ligand.

HiTrap Protein L columns are 1ml and 5ml columns prepacked with Capto L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
  • Prepacked HiTrap columns for easy screening and convenient process development
  • Suitable for small-scale purification
  • Columns are easily connected in a series for increased capacity
  • Designed for use with a syringe, peristaltic pump or chromatography system

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 × 5mL
Material Polypropylene (PP)
Content And Storage 2 to 8°C, 20% Ethanol
Column Type Affinity Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.