Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Protein L Prepacked Columns

Catalog No. 45002606 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
Diameter (English) Inner:
0.27 in.
0.62 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner
45-002-606 5 x 1mL 0.27 in.
45-002-605 1 x 5mL 0.62 in.
45-000-510 1 x 1mL 0.27 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-606 Supplier Cytiva Supplier No. 17547851
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs

  • Prepacked with Capto L affinity chromatography medium (resin)
  • Polypropylene column are easily connected in series for increased productivity
  • Easy screening and convenient process development
  • For use with syringe, peristaltic pump, or chromatography system
  • Maximum pressure: 5 bar
  • 1 and 5mL columns

BioProcess Media
  • Ligand: Recombinant protein L (E. coli), mammalian free
  • Matrix: Rigid, highly cross-linked agarose
  • Average particle size: 85μm
  • pH stability working range: 2 to 10

1mL columns

  • Column I.D.: 7mm
  • Flow velocity: <500cm/h3

5mL columns
  • Column I.D.: 16mm
  • Flow velocity: <500cm/h4

TRUSTED_SUSTAINABILITY

Specifications

Product Type HiTrap Protein L column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.78 in.
Height (Metric) 20mm
Flow Rate <500 cm/hr.
Material Polypropylene
Ligand Type Recombinant Protein L (E. coli), mammalian free
Max. Pressure 5 bar
pH Range 2 to 10
Particle Size 85μm
Support Type Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Affinity Column
For Use With (Application) Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.