Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ HiTrap™ Protein Select™ Column

Catalog No. 09928219 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-928-219 1 x 1 mL
09-928-217 1 x 5 mL
09-928-218 5 x 5 mL
09-928-220 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-928-219 Supplier Cytiva™ Supplier No. 17542151
Only null left
Add to Cart
Add to Cart

An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.

Cytiva™ Protein Select™ resin is an affinity chromatography resin for purifying any recombinant protein that does not have an affinity binding partner.

  • Simple affinity purification protocol for recombinant proteins.
  • One-step purification and tag cleavage without the need for proteases or other elution reagents.
  • Delivers a highly pure, native protein with no remaining tag amino acids after tag cleavage.
  • Can be processed under mild conditions, which means no exposure to conditions that cause protein aggregation or loss of biological activity.
  • Flexible, enabling the use in multiple expression systems.
TRUSTED_SUSTAINABILITY

Specifications

Type Column
Quantity 1 x 1 mL
Form Liquid
Column Type Pre-packed Column
Flow Rate 4 mL/min. Max
Volume (Metric) 1 x 1 mL
Diameter (Metric) 7 mm
Length (Metric) 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.