Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Protein A High Per
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17040303
Encompass
HiTrap(R) Protein A HP is a prepacked Protein A Sepharose(R) High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites serum and cell culture supernatants.HiTrap(R) Protein A HP is a prepacked ready to use column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column together with the medium provide fast simple and easy separations in a convenient formulation.
Catalog No. 50-290-8933