Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Streptavidin High
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17511201
Encompass
HiTrap(R) Streptavidin HP is a column prepacked with Streptavidin Sepharose(R) High Performance for purification of biotinylated proteins and biomolecules.
Catalog No. 50-290-7042
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.