Learn More
ACROBiosystems Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade C (16055)] GP120;HEK293;100UG;GP5
Supplier:  ACROBiosystems GP5V15224100UG
Encompass_Preferred
Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade C (16055)] GP120;100UG;HIV GP120 (HIV), His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 (Accession # ABL67444.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.0 kDa. The protein migrates as 85-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 2 μg/mL (100 μL/well) can bind HIV GP120 (HIV), His Tag (Cat. No. GP5-V15224) with a linear range of 0.5-63 ng/mL (QC tested).
Catalog No. 50-101-7916
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.