Learn More
CPC Scientific H-Ala-Val-Gly-Ile-Gly-Ala-Leu-Phe-Leu-Gly-Phe-Leu-Gly-Ala-Ala-Gly-Ser-Thr-Met-Gly-Ala-Arg-Ser-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV012A
Encompass
HIV envelope protein gp41 (519-541 (AVGIGALFLGFLGAAGSTMGARS-amide) destabilizes the lamellar structure of the membrane so that fusion can occur.ONE-LETTER SEQUENCE: AVGIGALFLGFLGAAGSTMGARS-NH2MOLECULAR FORMULA: C95H155N27O26S1MOLECULAR WEIGHT:2123.51STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [128631-86-7], RESEARCH AREA: AntimicrobialREFERENCES: J.L.Nieva et al., Lett. Pept. Sci., 4, 365 (1997); P.W.Mobley et al., Biochim. Biophys. Acta, 1418, 1 (1999); M.Kanyalkar et al., J. Pept. Sci., 10, 363 (2004)
Catalog No. 50-210-9551
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.