Learn More
CPC Scientific H-Gly-Leu-Asn-Lys-Ile-Val-Arg-Met-Tyr-Ser-Pro-Thr-Ser-Ile-Leu-Asp-Ile-Arg-OH (trifluoroacetate salt) 0.5MG
Supplier: CPC Scientific HIV016A
ONE-LETTER SEQUENCE: GLNKIVRMYSPTSILDIRMOLECULAR FORMULA: C92H158N26O26S1MOLECULAR WEIGHT:2076.49STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198636-98-5], RESEARCH AREA: AntimicrobialREFERENCES: M.J.E.Sternberg et al., FEBS Lett., 218, 231 (1987)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.