Learn More
Creative Biomart Recombinant Streptococcus pneumoniae type 7f Capsular Polysaccharide Antigen
Supplier:  Creative Biomart HIV264
Encompass_Preferred
Streptococcus pneumoniae type 7f capsular polysaccharide antigen is a vaccine that contains highly purified capsular polysaccharides from the invasive pneumococcal type 7f of Streptococcus pneumoniae.
Catalog No. 50-208-4642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.