Learn More
Creative Biomart Recombinant HIV-1 gp41 Long Antigen, b-galactosidase-tagged, HRP-Labled
Supplier:  Creative Biomart HIV268
Encompass_Preferred
The E. coli derived HRP Labeled protein contains HIV immunodominant regions from HIV-I gp41 and is fused to beta-galactosidase (114 kDa) at the N-terminus.
Catalog No. 50-208-4646
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.