Learn More
ACROBiosystems HIV-1 HXB2 GP41 50ug
Supplier:  ACROBiosystems GP1H51H350UG
Encompass_Preferred
HIV-1 (HXB2) GP41 Pre-hairpin intermediate Protein His Tag (GP1-H51H3) is expressed from E. coli cells. It contains AA Ala 533 - Leu 856 (Accession # P04578).
Catalog No. 50-253-4950
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.