Learn More
Creative Biomart Recombinant SARS Core Antigen
Supplier:  Creative Biomart HIV274
Encompass_Preferred
The E. coli derived 35 kDa recombinant protein contains Nucleocapsid protein 1-49, 192-220 amino acids immunodominant regions. The 2 regions are separated with 3 glycine residues.
Catalog No. 50-208-4652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.