Learn More
Creative Biomart Recombinant SARS Matrix Antigen
Supplier:  Creative Biomart HIV275
Encompass_Preferred
The E. coli derived 30 kDa recombinant protein contains the Matrix protein 182-216 amino acids immunodominant regions.
Catalog No. 50-208-4653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.