Learn More
Creative Biomart Recombinant HIV-1 p24 Antigen, His-tagged
Supplier:  Creative Biomart HIV281
Encompass_Preferred
HIV-1 p24 His Tag Recombinant produced in E. coli is a single, non-glycosylated polypeptide chain containing 188 amino acids (155-321 a.a.) and having a molecular mass of 21.2 kDa. The HIV-1 p24 is fu
Catalog No. 50-208-4659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.