Learn More
Creative Biomart Recombinant Rubella E1 Antigen, GST-tagged
Supplier:  Creative Biomart HIV288
Encompass_Preferred
The E. coli derived recombinant protein contains the Rubella Virus E1 regions, 157-176, 374-390, 213-239 amino acids. The protein is fused to a GST-tag.
Catalog No. 50-208-4666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.