Learn More
CPC Scientific H-Met-Asp-Pro-Val-Asp-Pro-Asn-Ile-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV019A
Encompass
MDPVDPNIE is an efficient dipeptidyl-peptidase IV (DPIV) inhibitor.ONE-LETTER SEQUENCE: MDPVDPNIEMOLECULAR FORMULA: C43H68N10O17S1MOLECULAR WEIGHT:1029.14STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [200203-20-9], RESEARCH AREA: AntimicrobialREFERENCES: S.Wrenger et al., J. Biol. Chem., 272, 30283 (1997); J.Mrestani-Klaus et al., J. Pept. Sci., 4, 400 (1998)
Catalog No. 50-210-9565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.