Learn More
CPC Scientific H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific TAT002B
Encompass
This is the most characterized fragment of the HIV transactivator protein (TAT). This arginine-rich TAT peptide penetrates plasma membrane directly, but not through endocytosis, and has been proven to promote the entry of nucleic acids into several cell types. ONE-LETTER SEQUENCE: YGRKKRRQRRRMOLECULAR FORMULA: C64H118N32O14MOLECULAR WEIGHT:1559.85STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [191936-91-1], SYNONYMS: HIV-1 tat Protein (47-57)RESEARCH AREA: AntimicrobialREFERENCES: C. Rudolph, et al.,JBC Papers in Press, M211891200, (2003); V.P. Torchilin, et al., PNAS, 100, 1972-1977, (2003); Wang, H. et al. J. Clin. Invest. 109, 1463 (2002); Ziegler et al. Biochem. 44, 138 (2005); Wender, PA. et al. Proc. Natl. Acad. Sci. USA 97, 13003 (2000)
Catalog No. 50-211-0540