Learn More
CPC Scientific H-Cys-Gly-Lys-Ile-Glu-Pro-Leu-Gly-Val-Ala-Pro-Thr-Lys-Ala-Lys-Arg-Arg-Val-Val-Gln-Arg-Glu-Lys-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV010B
Encompass
The sequence CGKIEPLGVAPTKAKRRVVQREKR corresponds to gp160 (a gp120-gp41 dimer) (486-508) N-terminally elongated with a cysteine residue.ONE-LETTER SEQUENCE: CGKIEPLGVAPTKAKRRVVQREKRMOLECULAR FORMULA: C117H211N41O31S1MOLECULAR WEIGHT:2720.29STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198636-94-1], RESEARCH AREA: Antimicrobial
Catalog No. 50-210-9548
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.