Learn More
Bon Opus Biosciences Recombinant HIV gp120 (C-8His) -50ug
SDP
Supplier:  Bon Opus Biosciences CP9350UG
Encompass_Preferred
Recombinant HIV 1 Envelope glycoprotein gp120 is produced by our Mammalian expression system and the target gene encoding Met1-Glu498 is expressed with a 8His tag at the C-terminus. Expression system: Human Cells. Accession# Q9DKG6. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CP93
Catalog No. 50-208-0386
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.