Learn More
Sino Biological HIV gp120 PAb 100UL 40259-T62-100
Supplier:  Sino Biological 40259T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1(group P, strain RBF168) gp120 (Catalog#40259-V08H1; ACY40659.1; Tyr38-Arg527). HIV-1(group P, strain RBF168) gp120 specific IgG was purified by HIV-1(group P, strain RBF168) gp120 affinity chromatography.
Catalog No. 50-240-9525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.