Learn More
Abcam HIV gp41 L Peptide, Synthetic, >95% Purity, 500 μg
Description
- Tested Applications: ELISA, WB
- Purity: >95%
- Form: Liquid
- Concentration: 1 mg/mL
- Protein/Peptide Type: Synthetic
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Shipping Condition | Blue Ice |
| Target Molecule | HIV gp41 |
| Substrate Properties | Peptide-based Substrate |
| Quantity | 500 μg |
| For Use With (Application) | ELISA, Western Blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.