Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- General Microbiology Quality Control Products
- ZeptoMetrix HIV Seroconversion Panels
ZeptoMetrix HIV Seroconversion Panels
Catalog No. 22157267
Shop All ZeptoMetrix Corporation Products

Change view
Click to view available options
Quantity:
3 x 1 mL
4 x 1 mL
5 x 1 mL
6 x 1 mL
7 x 1 mL
8 x 1 mL
9 x 1 mL
10 x 1 mL
11 x 1 mL
12 x 1 mL
13 x 1 mL
16 x 1 mL
17 x 1 mL
19 x 1 mL
22 x 1 mL
25 x 1 mL
29 x 1 mL
Specimen Type:
Donor No.: 1043487
Donor No.: 62216
Donor No.: 63331
Donor No.: 63753
Donor No.: 64469
Donor No.: 64578
Donor No.: 64954
Donor No.: 65376
Donor No.: 65389
Donor No.: 65404
Donor No.: 65522
Donor No.: 65685
Donor No.: 65790
Donor No.: 66048
Donor No.: 66575
Donor No.: 66632
Donor No.: 66686
Donor No.: 67485
Donor No.: 67512
Donor No.: 67706
Donor No.: 67860
Donor No.: 67996
Donor No.: 68205
Donor No.: 68262
Donor No.: 68582
Donor No.: 73695
Donor No.: 73698
Donor No.: 75018
Donor No.: 75062
Donor No.: 77600
Donor No.: 77840
Donor No.: 79065
32 product options available for selection
| Catalog No. | Quantity | Specimen Type |
|---|---|---|
| 22-157-267 | 9 x 1 mL | Donor No.: 64578 |
| 22-157-280 | 5 x 1 mL | Donor No.: 62216 |
| 22-157-286 | 4 x 1 mL | Donor No.: 77840 |
| 22-157-251 | 9 x 1 mL | Donor No.: 73695 |
| 22-157-283 | 5 x 1 mL | Donor No.: 79065 |
| 22-157-271 | 7 x 1 mL | Donor No.: 68205 |
| 22-157-279 | 13 x 1 mL | Donor No.: 66632 |
| 22-157-263 | 11 x 1 mL | Donor No.: 66575 |
| 22-157-269 | 12 x 1 mL | Donor No.: 64469 |
| 22-157-270 | 12 x 1 mL | Donor No.: 67996 |
| 22-157-260 | 7 x 1 mL | Donor No.: 65522 |
| 22-157-282 | 29 x 1 mL | Donor No.: 75018 |
| 22-157-289 | 6 x 1 mL | Donor No.: 1043487 |
| 22-157-259 | 7 x 1 mL | Donor No.: 65404 |
| 22-157-284 | 25 x 1 mL | Donor No.: 75062 |
| 22-157-287 | 4 x 1 mL | Donor No.: 77600 |
| 22-157-274 | 12 x 1 mL | Donor No.: 68262 |
| 22-157-268 | 22 x 1 mL | Donor No.: 67706 |
| 22-157-288 | 6 x 1 mL | Donor No.: 65376 |
| 22-157-262 | 10 x 1 mL | Donor No.: 65790 |
| 22-157-257 | 11 x 1 mL | Donor No.: 64954 |
| 22-157-276 | 19 x 1 mL | Donor No.: 66048 |
| 22-157-256 | 7 x 1 mL | Donor No.: 63331 |
| 22-157-281 | 10 x 1 mL | Donor No.: 63753 |
| 22-157-275 | 16 x 1 mL | Donor No.: 68582 |
| 22-157-264 | 3 x 1 mL | Donor No.: 65685 |
| 22-157-258 | 8 x 1 mL | Donor No.: 65389 |
| 22-157-273 | 7 x 1 mL | Donor No.: 67860 |
| 22-157-252 | 13 x 1 mL | Donor No.: 73698 |
| 22-157-266 | 17 x 1 mL | Donor No.: 67485 |
| 22-157-265 | 22 x 1 mL | Donor No.: 67512 |
| 22-157-278 | 16 x 1 mL | Donor No.: 66686 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Content And Storage | -20°C |
| Product Type | Seroconversion Reference Panel |
| Quantity | 9 x 1 mL |
| Specimen Type | Donor No.: 64578 |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?