Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 nef Monoclonal Antibody (3D12)
GREENER_CHOICE

Catalog No. MA171501
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171501 Supplier Invitrogen™ Supplier No. MA171501
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with HIV-1 subtypes B and C, BRU, IIIB, and HAN strains. It has been mapped to amino acids 35-50 (RDLEKHGAITSSNTAA - HXB2). This clone may also be referred to as 3D12 (01-002).

HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD 4 and MH4, important for AIDS virus infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 nef
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias HIV-1 Nef
Host Species Mouse
Immunogen Recombinant Nef protein of HIV-1 HXB2 purified from E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.