Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 p17 Monoclonal Antibody (17-1)
GREENER_CHOICE

Catalog No. PIMA518032 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518032 Supplier Invitrogen™ Supplier No. MA518032
  • $777.50
    $623.65 / Each of 1

    Save $153.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

.

HIV-1 Gag p17 is the matrix protein of AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitutes an essential element for the of AIDS virus particle construction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 p17
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 17-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias HIV1gp1
Host Species Mouse
Immunogen Bacterially expressed, hexahistidine amino-terminal tagged HIV-1 p17 Gag protein (clade B, HXB-3 isolate).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.